Learn More
Sino Biological Recombinant Human CD4 Protein (His & AVI Tag), Biotinylated, HPLC-verified
Supplier:  Sino Biological 10400H49HB100
Encompass_Preferred
A DNA sequence encoding the Human CD4 (NP_000607.1) (Met1-Pro396) was expressed with a C-terminal polyhistidine tag followed by an AVI tag. The expressed protein was biotinylated in vivo by the Biotin-Protein ligase (BirA enzyme) which is co-expressed.
Catalog No. 50-237-2328