Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Gibco™ Human CD4 (soluble) Recombinant Protein, PeproTech®

Catalog No. 11011100UG
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 μg
100 μg
250 μg
50 μg
500 μg
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11011100UG 100 μg
110-11-10UG 10 μg
11011250UG 250 μg
11011500UG 500 μg
110-11-50UG 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Catalog No. 11011100UG Supplier Gibco™ Supplier No. 11011100UG
Add to Cart
Add to Cart
This item is not returnable. View return policy

Recombinant Protein

PeproTech's CHO cell-derived Recombinant Human sCD4 is a monomeric glycoprotein of 371 amino acid residues, which correspond to the extracellular CD4 domain, and a calculated molecular weight of 41.3 kDa. As a result of glycosylation, Recombinant Human sCD4 migrates with an apparent molecular mass of approximately 45-55 kDa by SDS-PAGE Gel analysis, under reducing conditions. This product is shipped at ambient temperature. For storage, handling and reconstitution information, please see the lot-specific Certificate of Analysis

The CD4 antigen is involved in the recognition of MHC class II molecules and is a co-receptor for HIV. CD4 is primarily expressed in a subset of T-lymphocytes, also referred to as T helper cells, but may also be expressed by other cells in the immune system, such as monocytes, macrophages, and dendritic cells. At the tissue level, CD4 expression may be detected in thymus, lymph nodes, tonsils, and spleen, and also in specific regions of the brain, gut, and other non-lymphoid tissues. CD4 functions to initiate or augment the early phase of T-cell activation through its association with the T-cell receptor complex and protein tyrosine kinase, Lck. It may also function as an important mediator of direct neuronal damage in infectious and immune-mediated diseases of the central nervous system. Multiple alternatively spliced transcripts have been identified in this gene [RefSeq, July 2017].
TRUSTED_SUSTAINABILITY

Specifications

Accession Number P01730
For Use With (Application) Functional Assay
Formulation protein with no preservative
Gene ID (Entrez) 920
Molecular Weight (g/mol) 41.3 kDa
Name Human CD4 (soluble)
Quantity 100 μg
Source CHO cells
Endotoxin Concentration <1 EU/ μg
Gene Alias Activation B7-1 antigen; B7; B7.1; B7-1; BB1; B-lymphocyte activation antigen B7; CD28LG; CD28LG1; CD4; CD4 antigen; CD4 antigen (p55); CD4 antigen p55; Cd4 molecule; CD4 precursor; CD4 receptor; CD4, allele 1; cd4a; CD4mut; CD80; CD80 antigen (CD28 antigen ligand 1, B7-1 antigen); CD80 molecule; cell surface glycoprotein CD4; costimulatory factor CD80; costimulatory molecule variant IgV-CD80; CTLA-4 counter-receptor B7.1; fCD4; L3T4; LAB7; Leu-3; Ly-4; lymphocyte antigen CD4; lymphocyte antigen CD4 precursor; membrane protein; p55; T-cell differentiation antigen L3T4; T-cell surface antigen T4/Leu-3; T-cell surface glycoprotein CD4; T-cell surface glycoprotein CD4 precursor (T-cell surface antigen T4/Leu-3) (T-cell differentiation antigen L3T4); T-lymphocyte activation antigen CD80; W3/25; W3/25 antigen
Common Name CD4
Gene Symbol CD4
Biological Activity Determined by its ability to binds HeLa cells expressing cell-surface MHC-II molecules.
Conjugate Unconjugated
Recombinant Recombinant
Sequence KKVVLGKKGD TVELTCTASQ KKSIQFHWKN SNQIKILGNQ GSFLTKGPSK LNDRADSRRS LWDQGNFPLI IKNLKIEDSD TYICEVEDQK EEVQLLVFGL TANSDTHLLQ GQSLTLTLES PPGSSPSVQC RSPRGKNIQG GKTLSVSQLE LQDSGTWTCT VLQNQKKVEF KIDIVVLAFQ KASSIVYKKE GEQVEFSFPL AFTVEKLTGS GELWWQAERA SSSKSWITFD LKNKEVSVKR VTQDPKLQMG KKLPLHLTLP QALPQYAGSG NLTLALEAKT GKLHQEVNLV VMRATQLQKN LTCEVWGPTS PKLMLSLKLE NKEAKVSKRE KAVWVLNPEA GMWQCLLSDS GQVLLESNIK VLPTWSTPVQ P
Content And Storage -20°C
Expression System CHO cells
Form Lyophilized
Purity or Quality Grade ≥ 95% by SDS-PAGE gel and HPLC analyses.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.