Learn More
ACROBiosystems Human CD40 200tests
Catalog No. 502535394
Encompass_Preferred
Alexa Fluor 647-Labeled Human CD40 Protein Fc Tag (CD0-HA256) is produced via site-specific conjugation of AF647 to Human CD40 Protein Fc Tag under optimal conditions with a proprietary technology. Alexa Fluor 647-Labeled Human CD40 Protein Fc Tag is expressed from human 293 cells (HEK293). It contains AA Glu 21 - Arg 193 (Accession # P25942-1).
Catalog No. 50-253-5394 Supplier ACROBiosystems Supplier No. CD0HA256200TESTS
Alexa Fluor 647-Labeled Human CD40 Protein Fc Tag (CD0-HA256) is produced via site-specific conjugation of AF647 to Human CD40 Protein Fc Tag under optimal conditions with a proprietary technology. Alexa Fluor 647-Labeled Human CD40 Protein Fc Tag is expressed from human 293 cells (HEK293). It contains AA Glu 21 - Arg 193 (Accession # P25942-1).