Learn More
Sino Biological Anti-CD40/TNFRSF5 Antibody (APC), Mouse Monoclonal
Supplier:  Sino Biological 10774MM03A100
Encompass_Preferred
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human CD40 / TNFRSF5 (rh CD40 / TNFRSF5; Catalog#10774-H08H; NP_001241.1; Met1-Arg193) and conjugated with APC under optimum conditions, the unreacted APC was removed.
Catalog No. 50-240-8795