Learn More
Sino Biological Anti-CD40/TNFRSF5 Antibody, Mouse Monoclonal
Supplier:  Sino Biological 10774MM11100
Encompass_Preferred
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human CD40 / TNFRSF5 (rh CD40 / TNFRSF5; Catalog#10774-H08H; NP_001241.1; Met1-Arg193). The IgG fraction of the cell culture supernatant was purified by Protein A affinity chromatography.
Catalog No. 50-240-8674