Learn More
Sino Biological Anti-CD40/TNFRSF5 Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 10774T16100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Human CD40 / TNFRSF5 (rh CD40 / TNFRSF5; Catalog#10774-H08H; NP_001241.1; Met1-Arg193). CD40 / TNFRSF5 specific IgG was purified by Human CD40 / TNFRSF5 affinity chromatography.
Catalog No. 50-240-9047