Learn More
Sino Biological Recombinant Human CD40 Ligand/TNFSF5 Protein (ECD, Low Endotoxin, hFc Tag), HPLC-verified
Catalog No. 502992697
Encompass_Preferred
A DNA sequence encoding the Human CD40LG (NP_000065.1) (Met113-Leu261 ) was fused with the Fc region of human IgG1 at the N-terminus.
Catalog No. 50-299-2697 Supplier Sino Biological Supplier No. 10239H01HE50
A DNA sequence encoding the Human CD40LG (NP_000065.1) (Met113-Leu261 ) was fused with the Fc region of human IgG1 at the N-terminus.