Learn More
Sino Biological Recombinant Anti-CD40 Ligand/TNFSF5 Antibody, Rabbit Monoclonal
Supplier:  Sino Biological 10239R30120
Encompass_Preferred
This antibody was obtained from a rabbit immunized with purified, recombinant Human CD40 Ligand/CD40L/CD154 (rh CD40 Ligand/CD40L/CD154; Catalog#10239-H08E; NP_000065.1; Glu108-Leu261).
Catalog No. 50-240-8390