Learn More
Sino Biological Anti-CD44 Antibody (FITC), Mouse Monoclonal
Supplier:  Sino Biological 12211MM02F100
Encompass_Preferred
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human CD44 (rh CD44; Catalog#12211-H08H; P16070-1; Met 1-Pro 220) and conjugated with FITC under optimum conditions, the unreacted FITC was removed.
Catalog No. 50-255-4410