Learn More
Sino Biological Recombinant Anti-CD44 Antibody, Rabbit Monoclonal
Supplier:  Sino Biological 12211R065100
Encompass_Preferred
This antibody was obtained from a rabbit immunized with purified, recombinant Human CD44 (rh CD44; Catalog#12211-H08H; P16070-1; Met 1-Pro 220).
Catalog No. 50-255-4404