Learn More
Sino Biological Anti-CD44 Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 12211RP01100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Human CD44 (rh CD44; Catalog#12211-H08H; P16070-1; Met1-Pro220). Total IgG was purified by Protein A affinity chromatography.
Catalog No. 50-255-4403