Learn More
ACROBiosystems Human CD45 (26-482), His Tag (CD5-H52H9) is expressed from human 293 cells (HEK293). It contains AA Gln 26 - Lys 482 (Accession # P08575-8).
Catalog No. 502253239
Encompass_Preferred
CD45 is a receptor protein tyrosine phosphatase, also known as Ly-5 or leukocyte common antigen. CD45 mainly involves in the initiation of T cell receptor signaling by controlling the activation of the Src family protein-tyrosine kinases Lck and Fyn. CD45 deficiency causes in T- and B-lymphocyte dysfunction in the form of severe combined immune deficiency. It also takes a significant role in autoimmune diseases and cancer as well as in infectious diseases including fungal infections.
This item is not returnable. View return policy
Catalog No. 50-225-3239 Supplier ACROBiosystems Supplier No. CD5H52H9100UG
CD45 is a receptor protein tyrosine phosphatase, also known as Ly-5 or leukocyte common antigen. CD45 mainly involves in the initiation of T cell receptor signaling by controlling the activation of the Src family protein-tyrosine kinases Lck and Fyn. CD45 deficiency causes in T- and B-lymphocyte dysfunction in the form of severe combined immune deficiency. It also takes a significant role in autoimmune diseases and cancer as well as in infectious diseases including fungal infections.