Learn More
ACROBiosystems Human CD45 200ug
Supplier:  ACROBiosystems CD5H82E8200UG
Encompass_Preferred
Biotinylated Human CD45 / PTPRC Protein HisAvitag (CD5-H82E8) is expressed from human 293 cells (HEK293). It contains AA Gln 26-Lys 482 (Accession # P08575-8).
Catalog No. 50-253-5377