Learn More
ACROBiosystems Human CD45 25ug
Catalog No. 502535376
Encompass_Preferred
Biotinylated Human CD45 / PTPRC Protein HisAvitag (CD5-H82E8) is expressed from human 293 cells (HEK293). It contains AA Gln 26-Lys 482 (Accession # P08575-8).
Catalog No. 50-253-5376 Supplier ACROBiosystems Supplier No. CD5H82E825UG
Biotinylated Human CD45 / PTPRC Protein HisAvitag (CD5-H82E8) is expressed from human 293 cells (HEK293). It contains AA Gln 26-Lys 482 (Accession # P08575-8).