Learn More
ACROBiosystems Human CD45 / PTPRC protein, His Tag
Catalog No. 502054867
Encompass_Preferred
Recombinant Protein;1MG;Human CD45, His Tag (CD5-H52H8) is expressed from human 293 cells (HEK293). It contains AA Gln 26 - Lys 577 (Accession # P08575-3).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 62.6 kDa.
Catalog No. 50-205-4867 Supplier ACROBiosystems Supplier No. CD5H52H81MG
Recombinant Protein;1MG;Human CD45, His Tag (CD5-H52H8) is expressed from human 293 cells (HEK293). It contains AA Gln 26 - Lys 577 (Accession # P08575-3).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 62.6 kDa.