Learn More
Sino Biological Human CD46 MAb 100ul (Sino Biological)
Supplier:  Sino Biological 12239MM05100
Encompass_Preferred
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human CD46 (rh CD46; Catalog#12239-H08H; EAW93471.1; Met1-Asp328). The IgG fraction of the cell culture supernatant was purified by Protein A affinity chromatography.
Catalog No. 50-237-1963