Learn More
Sino Biological Anti-CD46 Antibody (FITC), Mouse Monoclonal
Supplier:  Sino Biological 12239MM05F25
Encompass_Preferred
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human CD46 (rh CD46; Catalog#12239-H08H; EAW93471.1; Met 1-Asp 328) and conjugated with FITC under optimum conditions, the unreacted FITC was removed.
Catalog No. 50-255-4391