Learn More
ACROBiosystems Human CD47 96tests
Catalog No. 502535101
Encompass_Preferred
CD47 is a transmembrane protein a cell surface glycoprotein molecule belonging to the immunoglobulin superfamily that binds to a variety of proteins including integrins thrombospondin 1 and signal regulatory protein-α (SIRPα). In recent years accumulating data have demonstrated that the CD47-SIRPαV2 axis is a key immune checkpoint in different cancers including hematological malignancies similar to PD-1/PD-L1 in solid tumors. CD47-SIRPαV2 blockade has become a next-generation immune checkpoint blocking strategy for various malignancies after PD-1/PD-L1.
Catalog No. 50-253-5101 Supplier ACROBiosystems Supplier No. EP16096TESTS
CD47 is a transmembrane protein a cell surface glycoprotein molecule belonging to the immunoglobulin superfamily that binds to a variety of proteins including integrins thrombospondin 1 and signal regulatory protein-α (SIRPα). In recent years accumulating data have demonstrated that the CD47-SIRPαV2 axis is a key immune checkpoint in different cancers including hematological malignancies similar to PD-1/PD-L1 in solid tumors. CD47-SIRPαV2 blockade has become a next-generation immune checkpoint blocking strategy for various malignancies after PD-1/PD-L1.