Learn More
ACROBiosystems CD47 [Biotinylated] : SIRP alpha Inhibitor Screening ELISA Kit
Catalog No. 502331110
Encompass_Preferred
CD47 is a transmembrane protein, a cell surface glycoprotein molecule belonging to the immunoglobulin superfamily that binds to a variety of proteins, including integrins, thrombospondin 1, and signal regulatory protein-α (SIRPα). In recent years, accumulating data have demonstrated that the CD47-SIRP-α axis is a key immune checkpoint in different cancers, including hematological malignancies, similar to PD-1/PD-L1 in solid tumors. Cd47-sirpα blockade has become a next-generation immune checkpoint blocking strategy for various malignancies after PD-1/PD-L1.
Catalog No. 50-233-1110 Supplier ACROBiosystems Supplier No. EP13896TESTS
CD47 is a transmembrane protein, a cell surface glycoprotein molecule belonging to the immunoglobulin superfamily that binds to a variety of proteins, including integrins, thrombospondin 1, and signal regulatory protein-α (SIRPα). In recent years, accumulating data have demonstrated that the CD47-SIRP-α axis is a key immune checkpoint in different cancers, including hematological malignancies, similar to PD-1/PD-L1 in solid tumors. Cd47-sirpα blockade has become a next-generation immune checkpoint blocking strategy for various malignancies after PD-1/PD-L1.