Learn More
Sino Biological Human CD47 MAb 100ul (Sino Biological)
Supplier:  Sino Biological 12283MM42100
Encompass_Preferred
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human CD47 (rh CD47; Catalog#12283-H08H; NP_942088.1; Met1-Pro139). The IgG fraction of the cell culture supernatant was purified by Protein A affinity chromatography.
Catalog No. 50-237-1400