Learn More
Sino Biological Recombinant Human CD47 Protein (His & AVI Tag), Biotinylated, HPLC-verified
Supplier:  Sino Biological 12283H27HB20
Encompass_Preferred
A DNA sequence encoding the human CD47 (NP_942088.1) (Gln19-Pro139) was expressed with a c-terminal polyhistidine tagged AVI tag at the C-terminus. The expressed protein was biotinylated in vivo by the Biotin-Protein ligase (BirA enzyme) which is co-expressed
Catalog No. 50-255-4383