Learn More
Sino Biological Recombinant Anti-CD5 Antibody (PerCP), Rabbit Monoclonal
Supplier:  Sino Biological 11027R039C100
Encompass_Preferred
This antibody was obtained from a rabbit immunized with purified, recombinant Human CD5 (rh CD5; Catalog#11027-H08H; NP_055022.2; Met1-Pro372) and conjugated with PerCP under optimum conditions, the unreacted PerCP was removed.
Catalog No. 50-241-2475