Learn More
Sino Biological Recombinant Human CD52 Protein (hFc & AVI Tag), Biotinylated
Supplier:  Sino Biological 15105H41HB100
Encompass_Preferred
A DNA sequence encoding the human CD52 (NP_001794.2) (Met1-Ser36) was expressed with a C-terminal Fc region of human IgG1 tag followed by an AVI tag. The expressed protein was biotinylated in vivo by the Biotin-Protein ligase (BirA enzyme) which is co-expressed.
Catalog No. 50-255-4110