Learn More
ACROBiosystems ACROBiosystems Human CD55 / DAF Protein, His Tag
Supplier:  ACROBiosystems CD5H52251MG
Encompass_Preferred
Recombinant Protein;1MG;Human CD55, His Tag (CD5-H5225) is expressed from human 293 cells (HEK293). It contains AA Asp 35 - Ser 353 (Accession # NP_000565.1).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 35.8 kDa. The protein migrates as 50-65 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.
Catalog No. 50-210-6561