Learn More
Sino Biological Anti-CD55/DAF Antibody (HRP), Mouse Monoclonal
Supplier:  Sino Biological 10101MM03H100
Encompass_Preferred
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human CD55 / DAF (rh CD55; Catalog#10101-H08H; NP_000565.1; Met 1-Ser 353). The IgG fraction of the cell culture supernatant was purified by Protein A affinity chromatography and conjugated with horseradish-peroxidase (HRP).
Catalog No. 50-241-1260