Learn More
Sino Biological Recombinant Anti-CD55/DAF Antibody, Rabbit Monoclonal
Supplier:  Sino Biological 10101R02820
Encompass_Preferred
This antibody was obtained from a rabbit immunized with purified, recombinant Human CD55 / DAF (rh CD55; Catalog#10101-H08H; NP_000565.1; Met 1-Ser 353).
Catalog No. 50-240-8801