Learn More
ACROBiosystems ACROBiosystems Human CD58 / LFA-3 Protein, His Tag
Catalog No. 502106700
Encompass_Preferred
Recombinant Protein;100UG;Human CD58, His Tag (LF3-H5225) is expressed from human 293 cells (HEK293). It contains AA Phe 29 - Arg 215 (Accession # AAH05930).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 22.3 kDa. The protein migrates as 30-55 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Human CD58, His Tag (Cat. No. LF3-H5225) at 10 μg/mL (100 μL/well) can bind Human CD2, Fc Tag (Cat. No. CD2-H5258) with a linear range of 5-78 ng/mL (QC tested).
Catalog No. 50-210-6700 Supplier ACROBiosystems Supplier No. LF3H5225100UG
Recombinant Protein;100UG;Human CD58, His Tag (LF3-H5225) is expressed from human 293 cells (HEK293). It contains AA Phe 29 - Arg 215 (Accession # AAH05930).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 22.3 kDa. The protein migrates as 30-55 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Human CD58, His Tag (Cat. No. LF3-H5225) at 10 μg/mL (100 μL/well) can bind Human CD2, Fc Tag (Cat. No. CD2-H5258) with a linear range of 5-78 ng/mL (QC tested).