Learn More
Sino Biological Anti-CD58/LFA-3 Antibody (PerCP), Mouse Monoclonal
Supplier:  Sino Biological 12409MM05C100
Encompass_Preferred
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human LFA-3 / CD58 (rh LFA-3 / CD58; Catalog#12409-H08H; AAH05930.1; Met1-Arg215) and conjugated with PerCP under optimum conditions, the unreacted PerCP was removed.
Catalog No. 50-240-9556