Learn More
Sino Biological Recombinant Anti-CD58/LFA-3 Antibody, Rabbit Monoclonal
Supplier:  Sino Biological 12409R083100
Encompass_Preferred
This antibody was obtained from a rabbit immunized with purified, recombinant Human LFA-3 / CD58 (rh LFA-3 / CD58; Catalog#12409-H08H; AAH05930.1; Met1-Arg215).
Catalog No. 50-240-9550