Learn More
Sino Biological Anti-CD58/LFA-3 Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 12409T50100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Human LFA-3 / CD58 (rh LFA-3 / CD58; Catalog#12409-H08H; AAH05930.1; Met1-Arg215). LFA-3 / CD58 specific IgG was purified by Human LFA-3 / CD58 affinity chromatography.
Catalog No. 50-241-1762