Learn More
Abcam Human CD59 ELISA Kit, 1 x 96 Tests
Catalog No. 502656708
Encompass_Preferred
Human CD59 ELISA Kit is a single-wash 90-min Simplestep used to quantify Human CD59 with a sensitivity of 4.9 pg/ml. The assay uses a simple mix-wash-read protocol with just one incubation and one wash step. - Colorimetric Sandwich ELISA - 450 nm readout : works on any standard plate reader - Design your own immunoassay: we also offer the conjugation-ready antibody pair
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-265-6708 Supplier Abcam Supplier No. AB2638931X96TESTS
Human CD59 ELISA Kit is a single-wash 90-min Simplestep used to quantify Human CD59 with a sensitivity of 4.9 pg/ml. The assay uses a simple mix-wash-read protocol with just one incubation and one wash step. - Colorimetric Sandwich ELISA - 450 nm readout : works on any standard plate reader - Design your own immunoassay: we also offer the conjugation-ready antibody pair
The product is subject to the following: Abcam Restricted Use Statement