Learn More
Sino Biological Recombinant Anti-CD59 Antibody, Rabbit Monoclonal
Supplier:  Sino Biological 12474R029100
Encompass_Preferred
This antibody was obtained from a rabbit immunized with purified, recombinant Human CD59 (rh CD59; Catalog#12474-H08H; P13987-1; Met 1-Glu 101).
Catalog No. 50-241-0501