Learn More
Sino Biological Human CD59 MAb 100ul (Sino Biological)
Supplier:  Sino Biological 12474R031100
Encompass_Preferred
This antibody was obtained from a rabbit immunized with purified, recombinant Human CD59 (rh CD59; Catalog#12474-H08H; P13987-1; Met1-Glu101).
Catalog No. 50-237-2034