Learn More
BIOHIPPO Human CD5L Microsample
SDP
Catalog No. 5000315125
Encompass
Human CD5L Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of CD5L in serum plasma tissue homogenates cell lysates and other biological fluids It provides a detection range of 0 16-10 ng/mL with a sensitivity of 0 053 ng/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Human CD5L Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Human CD5L Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Human CD5L biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color
Catalog No. 50-003-15125 Supplier BIOHIPPO Supplier No. BHE15210983
Human CD5L Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of CD5L in serum plasma tissue homogenates cell lysates and other biological fluids It provides a detection range of 0 16-10 ng/mL with a sensitivity of 0 053 ng/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Human CD5L Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Human CD5L Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Human CD5L biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color