Learn More
BIOHIPPO Human CD6 Microsample
SDP
Catalog No. 5000314333
Encompass
Human CD6 Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of CD6 in Serum plasma tissue homogenates and other biological fluids It provides a detection range of 31 25-2000 pg/mL with a sensitivity of 11 8 pg/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Human CD6 Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Human CD6 Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Human CD6 biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color
Catalog No. 50-003-14333 Supplier BIOHIPPO Supplier No. BHE15212438
Human CD6 Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of CD6 in Serum plasma tissue homogenates and other biological fluids It provides a detection range of 31 25-2000 pg/mL with a sensitivity of 11 8 pg/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Human CD6 Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Human CD6 Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Human CD6 biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color