Learn More
Raybiotech Inc Human CD163 ELISA Kit, 2-Pack
Supplier:  Raybiotech Inc ELHCD1632
Encompass_Preferred
RayBio Human CD163 ELISA Kit for cell culture supernatants, heparin and/or citrate treated plasma, and serum samples. EDTA plasma is not suitable for use in this assay due to its chelating property.
Catalog No. NC1702758