Learn More
Sino Biological Anti-CD63 Antibody (FITC), Mouse Monoclonal
Supplier:  Sino Biological 11271MM12F100
Encompass_Preferred
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human CD63 (rh CD63; Catalog#11271-H08H; P08962-1; Ala 103-Val 203) and conjugated with FITC under optimum conditions, the unreacted FITC was removed.
Catalog No. 50-241-2451