Learn More
Sino Biological Recombinant Anti-CD63 Antibody, Rabbit Monoclonal
Supplier:  Sino Biological 11271R001100
Encompass_Preferred
This antibody was obtained from a rabbit immunized with purified, recombinant Human CD63 / Tspan-30 / Tetraspanin-30 (rh CD63 / Tspan-30 / Tetraspanin-30; Catalog#11271-H08H; P08962-1; Ala103-Val203).
Catalog No. 50-241-0837