Learn More
Sino Biological Anti-CD63 Antibody, Mouse Monoclonal
Supplier:  Sino Biological 11271MM12100
Encompass_Preferred
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human CD63 / Tspan-30 / Tetraspanin-30 (rh CD63 / Tspan-30 / Tetraspanin-30; Catalog#11271-H08H; P08962-1; Ala103-Val203). The IgG fraction of the cell culture supernatant was purified by Protein A affinity chromatography.
Catalog No. 50-241-2456