Learn More
Sino Biological Recombinant Human CD63 Protein (His & AVI Tag), Biotinylated, HPLC-verified
Supplier:  Sino Biological 11271H49HB100
Encompass_Preferred
A DNA sequence encoding the Human CD63 (P08962-1)(Ala103-Val203) was expressed with a C-terminal polyhistidine tag followed by an AVI tag. The expressed protein was biotinylated in vivo by the Biotin-Protein ligase (BirA enzyme) which is co-expressed.
Catalog No. 50-255-4749