Learn More
Sino Biological Recombinant Human CD63 Protein (hFc Tag), HPLC-verified
Supplier:  Sino Biological 11271H02H1
Encompass_Preferred
A DNA sequence encoding the Human CD63 (P08962-1) (Ala103-Val203) was expressed with the Fc region of human IgG1 at the C-terminus.
Catalog No. 50-240-8577