Learn More
Abcam Human CD64 Antibody Pair - BSA and Azide free, 10 x 96 Tests
Catalog No. 502692942
Encompass_Preferred
Human CD64 Antibody Pair - BSA and Azide free is a kit containing recombinant capture and detector antibodies in a carrier-free formulation for the measurement of Human CD64.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-269-2942 Supplier Abcam Supplier No. AB30068310X96TESTS
Human CD64 Antibody Pair - BSA and Azide free is a kit containing recombinant capture and detector antibodies in a carrier-free formulation for the measurement of Human CD64.
The product is subject to the following: Abcam Restricted Use Statement