Learn More
Bon Opus Biosciences Recombinant Human CD68 (C-hFc) 50ug
SDP
Supplier:  Bon Opus Biosciences BP107650UG
Encompass_Preferred
Recombinant Human CD68 (C-hFc) is produced by Human Cells. The target gene encoding Asn22-Ser319 is expressed with a C-hFc tag. Accession# P34810. Formulation: Lyophilized from a 0.2 μm filtered solution of PBS, pH 7.4. Bon Opus Cat# BP1076
Catalog No. 50-217-2674