Learn More
Sino Biological Recombinant Anti-CD68 Antibody, Rabbit Monoclonal
Supplier:  Sino Biological 11192R179100
Encompass_Preferred
This antibody was obtained from a rabbit immunized with purified, recombinant Human CD68 (rh CD68; Catalog#11192-H08B1; P34810-1; Met1-Ser319).
Catalog No. 50-255-4794