Learn More
Sino Biological Anti-CD68 Antibody (FITC), Mouse Monoclonal
Supplier:  Sino Biological 11192MM11F25
Encompass_Preferred
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human CD68 (rh CD68; Catalog#11192-H08B1; P34810-1; Met1-Ser319). The IgG fraction of the cell culture supernatant was purified by Protein A affinity chromatography, and conjugated with FITC under optimum conditions, the unreacted FITC was removed.
Catalog No. 50-255-4799