Learn More
Sino Biological Recombinant Human CD68 Protein (ECD, hFc Tag)
Supplier:  Sino Biological 11192H02H100
Encompass_Preferred
A DNA sequence encoding the human CD68 (NP_001242.2) (Met1-Ser319) was expressed with the Fc region of human IgG1 at the C-terminus.
Catalog No. 50-255-4803