Learn More
Sino Biological Anti-CD69 Antibody (PerCP), Mouse Monoclonal
Supplier:  Sino Biological 11150MM06C100
Encompass_Preferred
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human CD69 (rh CD69; Catalog#11150-H08H; NP_001772.1; Ser62-Lys199) and conjugated with PerCP under optimum conditions, the unreacted PerCP was removed.
Catalog No. 50-240-8982