Learn More
Sino Biological Anti-CD69 Antibody, Mouse Monoclonal
Supplier:  Sino Biological 11150MM0520
Encompass_Preferred
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human CD69 / CLEC2C (rh CD69 / CLEC2C; Catalog#11150-H08H; NP_001772.1; Ser62-Lys199). The IgG fraction of the cell culture supernatant was purified by Protein A affinity chromatography.
Catalog No. 50-240-8693