Learn More
Sino Biological Human CD69 Matched ELISA Antibody Pair Set 15 Plates
Catalog No. 5000786758
Encompass_Preferred
The Human CD69 ELISA Pair Set is for the quantitative determination of Human CD69.The Sino Biological ELISA Pair Set is a solid phase sandwich ELISA (Enzyme-Linked Immunosorbent Assay). It utilizes a monoclonal antibody specific for Human CD69 coated on a 96-well plate. Standards and samples are added to the wells, and any Human CD69 present binds to the immobilized antibody. The wells are washed and a horseradish peroxidase conjugated mouse anti-Human CD69 monoclonal antibody is then added, producing an antibody-antigen-antibody "sandwich". The wells are again washed and TMB substrate solution is loaded, which produces color in proportion to the amount of Human CD69 present in the sample. To end the enzyme reaction, the stop solution is added and absorbances of the microwell are read at 450 nm.
Catalog No. 50-007-86758 Supplier Sino Biological Supplier No. SEK1115015
The Human CD69 ELISA Pair Set is for the quantitative determination of Human CD69.The Sino Biological ELISA Pair Set is a solid phase sandwich ELISA (Enzyme-Linked Immunosorbent Assay). It utilizes a monoclonal antibody specific for Human CD69 coated on a 96-well plate. Standards and samples are added to the wells, and any Human CD69 present binds to the immobilized antibody. The wells are washed and a horseradish peroxidase conjugated mouse anti-Human CD69 monoclonal antibody is then added, producing an antibody-antigen-antibody "sandwich". The wells are again washed and TMB substrate solution is loaded, which produces color in proportion to the amount of Human CD69 present in the sample. To end the enzyme reaction, the stop solution is added and absorbances of the microwell are read at 450 nm.