Learn More
Sino Biological Anti-CD69 Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 11150RP01100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Human CD69 (rh CD69; Catalog#11150-H08H; NP_001772.1; Ser 62-Lys 199). Total IgG was purified by Protein A affinity chromatography.
Catalog No. 50-241-0723